Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Science Vocabulary Bulletin Board Set
SDPSCI_ED

Catalog No. s02427
Click to view available options
Grade:
4 to 8
This item is not returnable. View return policy

Catalog No. S02427

Supplier: Carson Dellosa Publishing CD410029

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

An innovative, science-themed bulletin board set.

  • Suitable for grades 4 to 8
  • Teaches 48 important words from scientific disciplines including physics, astronomy, biology, geology and chemistry

Specifications

Grade 4 to 8
Format Print
Product Type Science Vocabulary
Subject Science
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.