Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scientific Device Protozoa Stain Control Slides
SDP

Catalog No. 23769502 Shop All Scientific Device Laboratory Inc. Products
Click to view available options
Quantity:
10/Pk.

Catalog No. 23-769-502

Supplier: Scientific Device 354

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Ideal for control of Trichrome or Hematoxylin staining procedures

  • Contains smear of mixture of commonly seen protozoa in polyvinyl alcohol
  • Available fixed with parasafe, copper or zinc

Specifications

Type Control Slide
Quantity 10/Pk.
For Use With (Application) For trichrome or hematoxylin staining procedures
Includes Smear of clinical protozoans in PVA, fixed with parasafe, copper/zinc
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.