Learn More
Eppendorf™ Lab Shaker Clamps
Unlock the potential of your experiments with Eppendorf's versatile range of shaker clamps. Securely fasten Erlenmeyer flasks, Thomson Ultra Yield™, Optimum Growth™, Tunair™, Corning™, and more to your universal platforms. Find the ideal clamp for every flask type.
$52.33 - $257.24
Catalog Number | Mfr. No. | Holds | Dimensions (L x W) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Holds | Dimensions (L x W) | Price | Quantity & Availability | ||||
14-284-41
![]() |
Eppendorf™
M11909004 |
25 mL Erlenmeyer Flasks | 23 x 22 mm |
Each for $52.33
|
|
||||
14-278-160
|
Eppendorf™
M11909000 |
50 mL Erlenmeyer Flasks | 51 x 26 mm |
Each for $54.04
|
![]() |
||||
14-278-161
|
Eppendorf™
M11909001 |
125 mL Erlenmeyer Flasks | 68 x 37 mm |
Each for $55.28
|
![]() |
||||
14-278-162
|
Eppendorf™
M11909002 |
250 mL Erlenmeyer Flasks | 91 x 47 mm |
Each for $85.08
|
|
||||
14-278-163
|
Eppendorf™
M11909003 |
500 mL Erlenmeyer Flasks | 102 x 48 mm |
Each for $96.41
|
|
||||
14-281-31
|
Eppendorf™
ACSB500S |
500 mL Media Bottles, 450 or 600 mL TPP Bioreactors | 92 x 108 mm |
Each for $186.32
|
|
||||
14-281-26
|
Eppendorf™
ACE1000S |
1 L Erlenmeyer Flasks | 130 x 64 mm |
Each for $123.66
|
|
||||
14-281-32
|
Eppendorf™
ACSB1000S |
1 L Media Bottles | 108 x 150 mm |
Each for $179.83
|
|
||||
14-278-001
|
Eppendorf™
M11909005 |
2 L Narrow Neck Erlenmeyer, Thomson or Corning Flasks | 165 x 112 mm |
Each for $173.70
|
|
||||
14-281-27
![]() |
Eppendorf™
ACE2000S |
2 L Wide Neck Erlenmeyer Flasks | 158 x 72 mm |
Each for $181.84
|
![]() |
||||
14-281-30
|
Eppendorf™
ACFE2800S |
2.8 L Fernbach Flasks | 203 x 73 mm |
Each for $233.99
|
|
||||
05-414-365
|
Eppendorf™
ACE3000S |
3 L Erlenmeyer Flasks | 189 x 112 mm |
Each for $155.41
|
|
||||
14-281-28
|
Eppendorf™
ACE4000S |
4 L Erlenmeyer Flasks | 210 x 99 mm |
Each for $206.74
|
![]() |
||||
05-414-366
|
Eppendorf™
ACE5000S |
5 L Erlenmeyer Flasks | 222 x 145 mm |
Each for $167.56
|
|
||||
14-281-29
|
Eppendorf™
ACE6000S |
6 L Erlenmeyer Flasks | 244 x 100 mm |
Each for $257.24
|
![]() |
||||
Description
- Stainless-steel construction (not spot welded or riveted) for added strength and safety.
- Clamps 2 L and above come with an extra girdle for added security when agitating flasks at high speeds.
- Comes complete with required screws, and are fitted with coiled wire springs for added stability at elevated speeds.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Clamp | |
Stainless Steel |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.