Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scigen VIP-Fixative Leak-Proof Specimen Containers Buffered Zinc 10% Formalin Filled

Catalog No. p-6629009 Shop All Scigen Inc Products
Change view
Click to view available options
Capacity (Metric):
120 mL, pre-filled 50%
180 mL, pre-filled 50%
1900 mL, pre-filled 50%
40 mL, pre-filled 50%
480 mL, pre-filled 50%
60 mL, pre-filled 50%
960 mL, pre-filled 50%
Material:
PET
Polypropylene
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Material
23-730-598 40 mL, pre-filled 50% Polypropylene
23-730-599 60 mL, pre-filled 50% Polypropylene
23-730-600 120 mL, pre-filled 50% Polypropylene
23-730-601 180 mL, pre-filled 50% Polypropylene
23-730-602 480 mL, pre-filled 50% Polypropylene
23-730-603 960 mL, pre-filled 50% Polypropylene
23-730-604 1900 mL, pre-filled 50% PET
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 23-730-598 Supplier Scigen Supplier No. 1030Z

Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier.

Safely transport, store and retrieve pathology specimens using Scigen's Forma-Vial™ system. Containers from 30 mL to 7.5 Liters, filled with buffered zinc formalin, OSHA compliant.

  • Forma-Vial™ specimen containers are uniquely designed to be leak proof and protect against biohazardous or hazardous material release to the environment Highly visible OSHA compliant labeling
  • Sizes range from 30mL specimen containers to 7.5 Liters operating room organ buckets
  • Container openings range from 50 mm to 300 mm
  • Filled with buffered zinc formalin to 50% of container’s capacity
  • Buffered zinc 10% formalin to enhance nuclear detail in immunohistochemistry and special staining techniques
  • Chloride, sulfate and mercury free
  • Nonsterile containers marked with a lot code and expiration date, 4 year shelf life
  • Formalin waste designed to be treated in bulk using Neutralex or treated individually using Neutralex tablets (patent pending).
  • Manufactured using ISO 13485 and FDA quality systems

Specifications

Product Type Specimen Container Vial
Closure Included Yes
Graduated Yes
Material Polypropylene
Sterility Non-sterile
Capacity (English) 1.4 oz., pre-filled 50%
Capacity (Metric) 40 mL, pre-filled 50%
Closure Color White
Closure Material Polyethylene
Closure Size 50 mm
Closure Type Valve Cap
Color Translucent
Description Specimen Container Pre-filled With Zinc Buffered 10% Formalin
Diameter (Metric) 44 mm
For Use With (Equipment) Histology/cytology specimen containers
Graduations 10 mL
Height (English) 1.6 in.
Height (Metric) 41.5 mm
Includes 50% Filled Zinc Buffered 10% Formalin
Label OSHA Compliant
No. per Case 98/Cs.
No. per Pack 98/Pk.
Product Line Forma-Vial™ Pre-filled Specimen Containers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.