Learn More
Scigen Neutralex™ Neutra-Pads™ Grossing Pads
Shop All Scigen Inc ProductsDescription
Neutra-Pads™ were created to absorb and neutralize liquids from large and small specimens during the grossing process. They also are ideal for controlling formalin spills in storage areas and on transportation carts—wherever these liquids must be retained in an enclosed environment.
Designed specifically for use with other Neutralex™ products for maximum effectiveness in treating formalin and aldehyde waste, Neutra-Pads™ have been treated with Neutralex™, which is the only California Environmental Protection Agency-certified solution on the market today.
These pads employ a specially devised backing system that keeps formalin contained and neutralized within the pad itself. The result is a cleaner, drier, and safer environment. Each 8 x 10 in. pad can hold up to 80 mL of liquid.
- Neutralizes formalin on-contact while absorbing hazardous liquids
- Highly absorbent polymeric fiber embedded with formalin neutralizer
- Eliminates hazardous vapors
- Blue dissection surface consists of a porous highly-resilient polyethylene copolymer cutting area
- Plastic backing prevents the escape of harmful liquids, keeping you grossing or storage area clean and dry
- Available in two sizes: 8 x 10 in. or 11 x 17 in.
- Each 8 x 10 in. pad absorbs up to 80 mL of formalin
- Each 11 x 17 in. pad absorbs up to 150 mL of formalin
- Manufactured using ISO 13485 quality system
Specifications
Specifications
| Color | Blue |
| Description | Grossing and Storage Pads Instantaneously Neutralize Formalin While Rapidly Absoring Harmful Liquids and Reducing Vapors, Keeps Area Clean and Dry |
| Capacity (English) | 0.02 gal. |
| Capacity (Metric) | 80 mL |
| Material | Pre-treated Formalin Neutralizer, Plastic-backed Absorbent Pad |
| Type | Neutra-Pads™ |
| For Use With (Application) | For formalin, grossing and storage safety |
| Packaging Quantity | 10 Pads/Cs. |
| Dimensions (L x W) | 8 x 10 in. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.