Learn More
Scigen Neutralex™ Volumetric Calibrated Reaction Container
Shop All Scigen Inc ProductsDescription
The Neutralex™ Aldehyde Reaction Container is part of the Neutralex™ franchise, a California EPA certified disposal system of formalin and aldehyde waste. The reaction container has a 2.5 gallon capacity, and it was designed to receive your spent formalin or aldehyde waste for both storage and treatment. Treatment is recommended, however, when 1 or 2 gallons of waste have accumulated. Each container includes a vapor-resistant cap to keep vapors inside the container.
NeutralexÒ products are certified by the California Environmental Protection Agency. The seal indicates that the formalin treatment process results in non-hazardous drain safe waste.
- Designed for use with Neutralex certified treatment technology
- Lightweight and easy to handle
- Thick wall HDPE 2.5 gal. (9.5 L) capacity with 3.5 in. (89 mm) wide mouth
- Vapor resistance waste collection and treatment container
- Designed for treatment of formalin, paraformaldehyde, glutaraldehyde and OPA solutions
Specifications
Specifications
| Color | Translucent |
| Description | Volumetric calibrated reaction container |
| Capacity (English) | 2.5 gal. |
| Capacity (Metric) | 9.5 L |
| Material | HDPE |
| Type | Reaction Container |
| For Use With (Application) | For neutralization of formalin, paraldehyde, glutaraldehyde and OPA waste |
| Includes | Vapor resistant screw cap |
| Packaging Quantity | 1 Ea. |
| Weight (English) | 4 lb. |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.