Learn More
Scigen Neutralex™ Certified Formalin Neutralizer Starter Kit
Shop All Scigen Inc ProductsDescription
Starter packs are a convenient way to employ safety in the laboratory. Protect employee health against hazardous vapors and formalin waste with a CA EPA approved neutralizer.
Starter packs include:
- 4 pouches of NEUTRALEX™
- 1 NEUTRA-WIPES™ container (60 wipes)
- 1 NEUTRA-FORM™
- 10 Pk. of 8 x 10 in. NEUTRA-PADS™
- 1 NEUTRALEX™ pH and Aldehyde Test Kit
- 1 Reaction Container
Neutralex™ treatment powder was created to treat and dispose of formalin and aldehyde waste with maximum effectiveness and convenience—by simply pouring them down the drain.
Designed specifically for use with other Scigen products, Neutralex™ is the only California EPA certified solution on the market today. As a result, it has become the industry standard for the safe treatment and disposal of formalin and aldehyde waste.
For replacement supplies, see cat. no. NEUTRALEX™ 23730561, Formalin test kit 23730564, NEUTRA-PADS™ 23730570, NEUTRA-FORM™ 23730569 and NEUTRA-WIPES™ 23730576.
Specifications
Specifications
| Certifications/Compliance | CalEPA Certified Polution Prevention Technology |
| Description | Environmental, health and safety kit provides formalin abatement |
| Type | Formalin Neutralizer |
| For Use With (Application) | Formalin safety and disposal |
| Includes | Four pouches of Neutralex™, one aldehyde test kit, one 2.5 gal. reaction container, 60 Neutra Wipes™, ten Pk. 8 x 10 in. Neutra Pads™, one container of Neutra Form™ |
| Packaging Quantity | 1 Ea. |
| Weight (English) | 19 lb. |
| Weight (Metric) | 8.6 kg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.