Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Scigen VIP-Fixative™ Non-precipitating, 10% Formalin, Neutral pH
Shop All Scigen Inc Products
Click to view available options
Capacity:
3.8 L
19 L
Description
- Introduced in 2001, VIP-Fixative has been marketed under several well-known brands worldwide for use in automatic tissue processors as a non-precipitating formalin fixative
- VIP-Fixative formula will not form solids (precipitate) in tissue processors which can eliminate the need for routine warm water flushing by preventing precipitate damage to the rotary valve.
- Clear, colorless solution free of particles, neutral buffered 10% formalin based, for use in histological applications of fixation and storage of tissue specimens, soluble is dehydrant reagents
- Improves cellular details and improves cell wall rigidity resulting in easier cutting and uniform sections
- Non-sterile containers marked with a lot code and expiration date, 4 year shelf life
- Formalin waste designed to be treated in bulk using Neutralex
- Manufactured using ISO 13485 and FDA quality systems
Specifications
Specifications
Capacity | 19 L |
Closure Type | Lined Cap with Quick Release Faucet |
Color | Clear, Water White |
Compatibility | Compatible with Aqueous and Alcoholic Solutions |
Content And Storage | Room temperature |
Concentration | Neutral Buffered 10% Formalin |
For Use With (Equipment) | All Tissue Processors |
Product Type | Formalin |
Solution Type | Formalin |
For Use With (Application) | Tissue processor, to prevent instrument damage from solids with no impact on specimen integrity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction