Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCIMP Antibody (NVL-07), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222349PEATT594
Description
SCIMP Monoclonal specifically detects SCIMP in Human samples. It is validated for Flow Cytometry.Specifications
SCIMP | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
SCIMP | |
Recombinant intracellular part of human SCIMP. | |
0.1 mL | |
388325.0 | |
Human | |
IgG2a |
Flow Cytometry | |
NVL-07 | |
Flow Cytometry | |
transmembrane protein C17orf87, C17orf87, chromosome 17 open reading frame 87, DTFT5783, FLJ32580, SLP adapter and CSK-interacting membrane protein, SLP adaptor and CSK interacting membrane protein, SLP65/SLP76, Csk-interacting membrane protein, UNQ5783 | |
Mouse | |
Protein A purified | |
Primary | |
The mouse monoclonal antibody NVL-07 recognizes intracellular part of human transmembrane adaptor SCIMP. This protein of 17 kDa predicted Mw migrates as a 22 kDa band on SDS PAGE. | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction