Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCIN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555034
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555034 Supplier Invitrogen™ Supplier No. PA555034
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RNLASITRIV EVDVDANSLN SNDVFVLKLP QNSGYIWVGK GASQEEEKGA EYVASVLKCK TLRIQEGEEP EEFWNSLGGK KDYQTSPLLE TQAEDHPPRL YGCSNK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.

The SCIN gene, also known as scinderin, encodes for a calcium-dependent actin filament-severing protein that is involved in the regulation of the cytoskeleton. It plays a significant role in actin filament dynamics, influencing cellular processes such as cell shape, motility, secretion, and division. SCIN has been identified as a novel oncogene, with its abnormal expression strongly correlated to various cancers, including glioma and uterine corpus endometrial carcinoma. Specific mutations like S673F and S381Y in SCIN are linked with poor prognosis and are potential targets for cancer diagnosis, prognosis, and treatment strategies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCIN
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCIN
Gene Accession No. Q9Y6U3
Gene Alias Adseverin; AW545522; Gelsolin-like protein; KIAA1905; SCIN; Scind; Scinderin
Gene Symbols SCIN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCIN. Recombinant protein control fragment (Product #RP-92602).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85477
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.