Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCIN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA522243
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (92%), Rat (92%), Pig (94%), Chicken (83%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Specifications
SCIN | |
Polyclonal | |
Unconjugated | |
SCIN | |
Adseverin; AW545522; Gelsolin-like protein; KIAA1905; SCIN; Scind; Scinderin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
85477 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y6U3 | |
SCIN | |
Recombinant fragment corresponding to a region within amino acids 281 and 585 of SCIN (Uniprot ID#Q9Y6U3). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction