Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Globe Scientific Scintillation Vials
Click to view available options
Volume (Metric):
4 mL
6.5 mL
20 mL
Height (Metric):
53 mm
57 mm
61.4 mm
Diameter (Metric) Outer:
13.8 mm
16 mm
26.5 mm
Description
20mL Scintillation Vial:
- Scintillation counting, gamma counting, chromatography, sample storage and culturing
- Because of its leak-tight cap it is also commonly used to store and transport histological specimens in 10% formalin
- Provides high degree of resistance to organic solvents and sufficient translucence so that reagent levels can be easily seen
- Unique thread design of closure permits vial to be sealed tight with easy 1/4 turn and with minimum pressure
6.5mL Scintillation Vial:
- Securely sealed by simply snapping caps on
- Cap is conveniently removed with easy 1/4 turn to left
4mL Polyethylene Scintillation Vial:
- Uses full turn polyethylene screw cap
- Most economical size to use because it requires least amount of solvent
Specifications
Specifications
| Diameter (Metric) Outer | 26.5 mm |
| Material | HDPE |
| Sterility | Non-sterile |
| Volume (Metric) | 20 mL |
| No. per Case | 1000 |
| Closure Material | Polyethylene |
| Closure Type | Screw Cap |
| Height (Metric) | 61.4 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction