Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SCIO 469 hydrochloride
GSA_VAGREENER_CHOICESDP

Catalog No. 352850 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-285-0 50 mg
35-281-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-285-0 Supplier R&D Systems™ Supplier No. 3528/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective p38 MAPK inhibitor

SCIO 469 hydrochloride is a selective, ATP-competitive p38 inhibitor (IC50 = 9 nM for p38α in vitro). Displays approximately 10-fold selectivity for p38α over p38β and 2000-fold selectivity for p38α over 20 other kinases. Reduces p38α phosphorylation in multiple myeloma cells in vitro and in vivo; activity results in decreased tumor burden and angiogenesis in murine models of multiple myeloma. Also enhances bortezomib -induced cytotoxicity against multiple myeloma cells.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 309913-83-5
Molecular Formula C27H30ClFN4O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 549.46
Purity >98%
Molecular Weight (g/mol) 513.01
Chemical Name or Material SCIO 469 hydrochloride
Recommended Storage Desiccate at +4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.