Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106744
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106744 Supplier Invitrogen™ Supplier No. PA5106744
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SHC2.

Src homology 2 (SH2) domains bind specifically to tyrosine-phosphorylated proteins that temporally participate in signal transduction events. Shc-like protein (Sck) is a neuronal adaptor protein that contains an N-terminal PTB (phosphotyrosine binding) domain, a collagen homology (CH) domain, and a conserved C-terminal SH2 domain. Human Sck transcripts are present at high levels in liver, pancreas, prostate and ovary. In vascular endothelial cells, Sck participates in VEGF-induced signal transduction. Treatment of human umbilical vein endothelial (HUVEC) cells with VEGF induces recruitment of Sck to tyrosine-1175 of the kinase insert domain-containing receptor (KDR) and enhances Sck tyrosine phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCK
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SHC2
Gene Accession No. O70142, P98077
Gene Alias 6720466E06; neuronal Shc adaptor homolog; protein Sck; Protein Sli; SCK; SH2 domain protein C2; SHC (Src homology 2 domain containing) transforming protein 2; SHC adaptor protein 2; SHC2; ShcB; SHC-transforming protein 2; SHC-transforming protein B; SLI; src homology 2 domain-containing transforming protein C2; src homology 2 domain-containing-transforming protein C2
Gene Symbols SHC2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SHC2(Accession P98077), corresponding to amino acid residues A297-P347.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25759, 314612
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.