Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCL/Tal1 Antibody (BTL73), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308616R
Description
SCL/Tal1 Monoclonal antibody specifically detects SCL/Tal1 in Human samples. It is validated for Western Blot, ELISASpecifications
SCL/Tal1 | |
Monoclonal | |
DyLight 550 | |
BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5tal-1 product | |
Recombinant human full-length TAL1 protein (Uniprot: P17542) | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
BTL73 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
6886 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction