Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sclerostin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980746
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0746 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0746 Supplier Bioss Supplier No. BS10200R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Negative regulator of bone growth that acts through inhibition of Wnt signaling and bone formation.

Specifications

Antigen Sclerostin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SOST
Gene Accession No. Q9BQB4
Gene Alias 5430411E23Rik; CDD; DAND6; sclerosteosis; Sclerostin; Sost; SOST1; UNQ2976/PRO7455/PRO7476; VBCH
Gene Symbols SOST
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Sclerostin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50964
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.