Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCLY Monoclonal Antibody (3B2)
GREENER_CHOICE

Catalog No. PIMA521953
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521953 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521953 Supplier Invitrogen™ Supplier No. MA521953
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: NSQFPGTQRL PNTCNFSIRG PRLQGHVVLA QCRVLMASVG AACHSDHGDQ PSPVLLSYGV PFDVARNALR LSVGRSTTRA EVDLVVQDLK QAVAQLEDQ.

The SCLY gene encodes selenocysteine lyase, an enzyme involved in selenium metabolism, which plays a critical role in the recycling of selenocysteine from selenoproteins. SCLY catalyzes the conversion of selenocysteine into alanine, releasing selenium for reuse in the biosynthesis of new selenoproteins. It is chiefly active in the liver and kidneys, contributing to maintaining selenium homeostasis, which is vital for numerous biological functions due to selenium's role in antioxidant defense and thyroid hormone metabolism. Alterations in SCLY function or expression can affect selenium levels in the body and are associated with disorders related to oxidative stress and metabolic imbalances. Given the importance of selenium in cellular protection and immune function, SCLY is also of interest in studies focusing on dietary supplementation and disease prevention strategies, particularly in conditions like cancer and cardiovascular diseases where oxidative damage is prevalent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCLY
Applications ELISA, Immunoprecipitation
Classification Monoclonal
Clone 3B2
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Scly
Gene Accession No. Q96I15
Gene Alias 9830169H08; A930015N15Rik; hSCL; mSCL; SCL; SCLY; Scly1; Scly2; Selenocysteine lyase; selenocysteine lyase; Selenocysteine reductase; selenocysteine reductase
Gene Symbols Scly
Host Species Mouse
Immunogen SCLY (NP_057594, 346 a.a. ~ 444 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51540
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.