Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCLY Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563172
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563172 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563172 Supplier Invitrogen™ Supplier No. PA563172
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LYPMLFGGGQ ERNFRPGTEN TPMIAGLGKA AELVTQNCEA YEAHMRDVRD YLEERLEAEF GQKRIHLNS Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.

The SCLY gene encodes selenocysteine lyase, an enzyme involved in selenium metabolism, which plays a critical role in the recycling of selenocysteine from selenoproteins. SCLY catalyzes the conversion of selenocysteine into alanine, releasing selenium for reuse in the biosynthesis of new selenoproteins. It is chiefly active in the liver and kidneys, contributing to maintaining selenium homeostasis, which is vital for numerous biological functions due to selenium's role in antioxidant defense and thyroid hormone metabolism. Alterations in SCLY function or expression can affect selenium levels in the body and are associated with disorders related to oxidative stress and metabolic imbalances. Given the importance of selenium in cellular protection and immune function, SCLY is also of interest in studies focusing on dietary supplementation and disease prevention strategies, particularly in conditions like cancer and cardiovascular diseases where oxidative damage is prevalent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCLY
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Scly
Gene Accession No. Q96I15
Gene Alias 9830169H08; A930015N15Rik; hSCL; mSCL; SCL; SCLY; Scly1; Scly2; Selenocysteine lyase; selenocysteine lyase; Selenocysteine reductase; selenocysteine reductase
Gene Symbols Scly
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCLY. Recombinant protein control fragment (Product #RP-103918).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51540
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.