Learn More
SCLY Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 106671AP150UL

Description
This antibody detects the N-terminal of the protein.
Selenocysteine lyase (SCLY; EC 4.4.1.16) catalyzes the pyridoxal 5-prime phosphate-dependent conversion of L-selenocysteine to L-alanine and elemental selenium (Mihara et al., 2000).Specifications
SCLY | |
Polyclonal | |
Unconjugated | |
SCLY | |
hSCL, SCL, SCLY, selenocysteine lyase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
363285, 50880, 51540 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q68FT9, Q96I15, Q9JLI6 | |
Scly | |
SCLY Fusion Protein Ag0972 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.