Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCMH1 Antibody (OTI5E6), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274023V
Description
SCMH1 Monoclonal specifically detects SCMH1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SCMH1 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SCMH1 | |
Recombinant protein fragment corresponding to amino acids 1-309 of human SCMH1 (NP_036368) produced in E.coli. | |
0.1 mL | |
22955.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI5E6 | |
Western Blot, Immunohistochemistry | |
polycomb protein SCMH1, Scml3, Sex comb on midleg homolog 1, sex comb on midleg homolog 1 (Drosophila) | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction