Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCMH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530454
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530454 Supplier Invitrogen™ Supplier No. PA530454
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, IMR32, Mouse brain. Predicted reactivity: Mouse (85%), Rat (85%), Bovine (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Associates with Polycomb group (PcG) multiprotein complexes; the complex class is required to maintain the transcriptionally repressive state of some genes. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCMH1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene SCMH1
Gene Accession No. Q8K214, Q96GD3
Gene Alias AI315320; AI851618; Polycomb protein SCMH1; scmh1; Scml1; Scml3; sex comb on midleg 1; sex comb on midleg homolog 1; sex comb on midleg homolog 1 (Drosophila); sex comb on midleg-like 1; sex comb on midleg-like 3
Gene Symbols SCMH1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human SCMH1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22955, 29871
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.