Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCML1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584495
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584495 Supplier Invitrogen™ Supplier No. PA584495
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SNSSSEIDVQ EPNIVSDASC NTEEQLKTVD DVLIHCQVIY DALQNLDKKI DVIRRKVSKI QRFHVRSL Highest antigen sequence indentity to the following orthologs: Mouse - 28%, Rat - 30%.

Putative Polycomb group (PcG) protein. PcG proteins act by forming multiprotein complexes, which are required to maintain the transcriptionally repressive state of homeotic genes throughout development. May be involved in spermatogenesis during sexual maturation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCML1
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Scml1
Gene Accession No. Q9UN30
Gene Alias Scm polycomb group protein like 1; SCML1; sex comb on midleg like 1; sex comb on midleg-like 1 (Drosophila); sex comb on midleg-like protein 1
Gene Symbols Scml1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCML1. Recombinant protein control fragment (Product #RP-105184).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6322
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.