Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCML2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584666
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584666 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584666 Supplier Invitrogen™ Supplier No. PA584666
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SRSVPGTTSS PLVGDISPKS SPHEVKFQMQ RKSEAPSYIA VPDPSVLKQG FSKDPSTWSV DEVIQFMKHT DPQISGPLAD LFR Highest antigen sequence indentity to the following orthologs: Rat - 58%, Mouse - 57%.

SCML2 is a putative component of a Polycomb group (PcG) multiprotein complex involved in represing transcription of homeotic genes during development. It mediates chromatin remodeling and DNA dependent negative regulation of transcription by binding to methylated lysine residues in histone tails.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCML2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCML2
Gene Accession No. Q9UQR0
Gene Alias 4932420G07Rik; Scm polycomb group protein like 2; SCML2; sex comb on midleg-like 2 (Drosophila); sex comb on midleg-like protein 2
Gene Symbols SCML2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCML2. Recombinant protein control fragment (Product #RP-107363).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10389
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.