Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCML2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104133
Description
Antibody detects endogenous levels of total SCML2.
SCML2 is a putative component of a Polycomb group (PcG) multiprotein complex involved in represing transcription of homeotic genes during development. It mediates chromatin remodeling and DNA dependent negative regulation of transcription by binding to methylated lysine residues in histone tails.
Specifications
SCML2 | |
Polyclonal | |
Unconjugated | |
SCML2 | |
4932420G07Rik; Scm polycomb group protein like 2; SCML2; sex comb on midleg-like 2 (Drosophila); sex comb on midleg-like protein 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10389, 107815 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9UQR0 | |
SCML2 | |
A synthesized peptide derived from human SCML2(Accession Q9UQR0), corresponding to amino acid residues F448-R498. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction