Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCML4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566812
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566812 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566812 Supplier Invitrogen™ Supplier No. PA566812
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CSASEKVQEKE EGRMESVKTV TTEEYLVNPV GMNRYSVDTS ASTFNHRGSL HPSSSLYCKR QNSGD Highest antigen sequence identity to the following orthologs - mouse 56%, rat 65%.

Putative Polycomb group (PcG) protein. PcG proteins act by forming multiprotein complexes, which are required to maintain the transcriptionally repressive state of homeotic genes throughout development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCML4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCML4
Gene Accession No. Q8N228
Gene Alias 9330161D17; dJ47M23.1; mFLJ00197; Scml4; sex comb on midleg-like 4 (Drosophila); sex comb on midleg-like protein 4
Gene Symbols SCML4
Host Species Rabbit
Immunogen Recombinant Human SCML4. Recombinant protein control fragment (Product #RP-107470).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256380
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.