Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCN10A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113034
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113034 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113034 Supplier Invitrogen™ Supplier No. PA5113034
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCN10A Polyclonal Antibody for ICC/IF, ELISA

Scn10a is a tetrodotoxin-resistant voltage-gated sodium channel alpha subunit. Epithelial sodium channels are amiloride-sensitive members of the Degenerin/epithelial sodium channel (Deg/ENaC) superfamily of ion channels. Scn10a mediates the voltage-dependent sodium ion permeability of excitable membranes. Assuming opened or closed conformations in response to the voltage difference across the membrane, Scn10a forms a sodium-selective channel through which sodium ions may pass in accordance with their electrochemical gradient. Scn10a also plays a role in neuropathic pain mechanisms. Structurally, members of Deg/ENaC superfamily of ion channels share organizational similarity in that they all possess two short intracellular amino and carboxyl termini, two short membrane spanning segments, and a large extracellular loop with a conserved cysteine-rich region. There are three homologous isoforms of the ENaC (alpha, beta, and gamma) protein. ENaC in the kidney, lung, and colon plays an essential role in trans-epithelial sodium and fluid balance. ENaC also mediates aldosterone-dependent sodium reabsorption in the distal nephron of the kidney, thus regulating blood pressure. ENaC is thought to be regulated, in part, through association with the cystic fibrosis transmembrane conductance regulator (CFTR) chloride ion channel. Gain-of-function mutations in beta- or gamma-ENaC can cause severe arterial hypertension (Liddels syndrome) and loss-of-function mutations in alpha- or beta-ENaC causes pseudohypoaldosteronism (PHA-1). Diseases associated with SCN10A include Episodic Pain Syndrome, Familial, 2 and Sodium Channelopathy-Related Small Fiber Neuropathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCN10A
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Scn10a
Gene Accession No. Q9Y5Y9
Gene Alias FEPS2; hPN3; mPN3; Na(V)1.8; Nav1.8; peripheral nerve sodium channel 3; PN3; Scn10a; Sensory neuron sodium channel; SNS; Sodium channel protein type 10 subunit alpha; sodium channel protein type X subunit alpha; sodium channel type X alpha polypeptide; sodium channel voltage-gated type X alpha polypeptide; sodium channel, voltage gated, type X alpha subunit; sodium channel, voltage-gated, type 10, alpha polypeptide; sodium channel, voltage-gated, type X, alpha; sodium channel, voltage-gated, type X, alpha polypeptide; sodium channel, voltage-gated, type X, alpha subunit; sodium voltage-gated channel alpha subunit 10; voltage gated sodium channel; voltage-gated sodium channel subunit alpha Nav1.8
Gene Symbols Scn10a
Host Species Rabbit
Immunogen Recombinant Human Sodium channel protein type 10 subunit alpha protein (992-1099AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6336
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.