Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCN11A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114378
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114378 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114378 Supplier Invitrogen™ Supplier No. PA5114378
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human PC-3 whole cell, rat brain tissue, rat kidney tissue, rat C6 whole cell, mouse spleen tissue, mouse brain tissue, mouse kidney tissue. IHC: rat spleen tissue, mouse spleen tissue. Flow: U87 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Epithelial sodium channels are amiloride-sensitive members of the Degenerin/epithelial sodium channel (Deg/ENaC) superfamily of ion channels. Members of this superfamily of ion channels share organizational similarity in that they all possess two short intracellular amino and carboxyl termini, two short membrane spanning segments, and a large extracellular loop with a conserved cysteine-rich region. There are three homologous isoforms of the ENaC (alpha, beta, and gamma) protein. ENaC in the kidney, lung, and colon plays an essential role in trans-epithelial sodium and fluid balance. ENaC also mediates aldosterone-dependent sodium reabsorption in the distal nephron of the kidney, thus regulating blood pressure. ENaC is thought to be regulated, in part, through association with the cystic fibrosis transmembrane conductance regulator (CFTR) chloride ion channel. Gain-of-function mutations in beta- or gamma-ENaC can cause severe arterial hypertension (Liddels syndrome) and loss-of-function mutations in alpha- or beta-ENaC causes pseudohypoaldosteronism (PHA-1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCN11A
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene SCN11A
Gene Accession No. O88457, Q9R053, Q9UI33
Gene Alias FEPS3; hNaN; HSAN7; Nan; NaT; NaV1.9; NSS2; Peripheral nerve sodium channel 5; PN5; Scn11a; SCN12A; sensory neuron sodium channel 2; SNS2; SNS-2; Sodium channel protein type 11 subunit alpha; sodium channel protein type XI subunit alpha; sodium channel voltage-gated type 11 alpha polypeptide; sodium channel voltage-gated type XI alpha polypeptide; sodium channel, voltage gated, type XI alpha subunit; sodium channel, voltage-gated, type 11, alpha polypeptide; sodium channel, voltage-gated, type XI, alpha; sodium channel, voltage-gated, type XI, alpha polypeptide; sodium channel, voltage-gated, type XI, alpha subunit; sodium channel, voltage-gated, type XII, alpha polypeptide; sodium channel, voltage-gated, type11, alpha polypeptide; sodium voltage-gated channel alpha subunit 11; voltage-gated sodium channel NAV1.9b; voltage-gated sodium channel subunit alpha Nav1.9
Gene Symbols SCN11A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence of human SCN11A (MDDRCYPVIFPDERNFRPFTSDSLAAIEKRIAIQKEKKK).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11280, 24046, 29701
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.