Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCN1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115619
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115619 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115619 Supplier Invitrogen™ Supplier No. PA5115619
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Nav1.1.

Epithelial sodium channels are amiloride-sensitive members of the Degenerin/epithelial sodium channel (Deg/ENaC) superfamily of ion channels. Members of this superfamily of ion channels share organizational similarity in that they all possess two short intracellular amino and carboxyl termini, two short membrane spanning segments, and a large extracellular loop with a conserved cysteine-rich region. There are three homologous isoforms of the ENaC (alpha, beta, and gamma) protein. ENaC in the kidney, lung, and colon plays an essential role in trans-epithelial sodium and fluid balance. ENaC also mediates aldosterone-dependent sodium reabsorption in the distal nephron of the kidney, thus regulating blood pressure. ENaC is thought to be regulated, in part, through association with the cystic fibrosis transmembrane conductance regulator (CFTR) chloride ion channel. Gain-of-function mutations in beta- or gamma-ENaC can cause severe arterial hypertension (Liddels syndrome) and loss-of-function mutations in alpha- or beta-ENaC causes pseudohypoaldosteronism (PHA-1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCN1A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Scn1a
Gene Accession No. A2APX8, P04774, P35498
Gene Alias 3-Feb; B230332M13; BFIC3; BFIS3; BFNIS; brain III voltage-gated sodium channel; Brain Type I; cardiac tetrodotoxin-insensitive voltage-dependent sodium channel alpha subunit; CDCD2; CERIII; CIAT; CIN5; CMD1E; CMPD2; CMS16; CTC-264K15.6; EIEE11; EIEE13; EIEE6; ETHA; FEB3; FEB3A; FEB3B; FEPS2; FEPS3; FHM3; GEFS+2; GEFSP2; GEFSP7; HB1; HB2; HBA; HBBD; HBSC I; HBSC II; HBSCI; HBSCII; HH1; hNa6/Scn8a voltage-gated sodium channel; hNaN; hNE-Na; HOKPP2; hPN3; HSAN2D; HSAN7; HYKPP; HYPP; ICCD; IVF; KIAA1356; LQT3; MED; Na(v)1.2; Na(V)1.4; NAC1; NAC1A; NAC2; NAC3; NaCh6; NaN; Nav1.1; Nav1.2; Nav1.3; Nav1.4; Nav1.5; Nav1.6; Nav1.7; Nav1.8; NAV1.9; NENA; NE-NA; Neuroendocrine sodium channel; peripheral nerve sodium channel 3; Peripheral nerve sodium channel 5; peripheral sodium channel 1; PFHB1; PN1; PN3; PN4; PN5; RBI; SCN1; SCN10A; SCN11A; SCN12A; Scn1a; SCN2A; SCN2A1; SCN2A2; SCN3A; SCN4A; SCN5A; SCN8A; SCN9A; Sensory neuron sodium channel 2; SFNP; skeletal muscle sodium channel alpha subunit; skeletal muscle voltage-dependent sodium channel type IV alpha subunit; SkM1; SMEI; SNS; SNS2; SNS-2; Sodium channel protein brain I subunit alpha; sodium channel protein brain II subunit alpha; Sodium channel protein brain III subunit alpha; sodium channel protein cardiac muscle subunit alpha; Sodium channel protein skeletal muscle subunit alpha; sodium channel protein type 1 subunit alpha; Sodium channel protein type 10 subunit alpha; sodium channel protein type 11 subunit alpha; Sodium channel protein type 2 subunit alpha; sodium channel protein type 3 subunit alpha; Sodium channel protein type 4 subunit alpha; sodium channel protein type 5 subunit alpha; Sodium channel protein type 8 subunit alpha; sodium channel protein type 9 subunit alpha; Sodium channel protein type I subunit alpha; sodium channel protein type II subunit alpha; Sodium channel protein type III subunit alpha; sodium channel protein type IV subunit alpha; Sodium channel protein type IX subunit alpha; Sodium channel protein type V subunit alpha; Sodium channel protein type VIII subunit alpha; Sodium channel protein type X subunit alpha; sodium channel protein type XI subunit alpha; sodium channel protein, brain I alpha subunit; sodium channel protein, brain I subunit alpha; sodium channel protein, brain type 2 alpha subunit; sodium channel voltage gated type 1 alpha subunit; sodium channel voltage-gated type I alpha polypeptide; sodium channel, voltage gated, type I alpha subunit; sodium channel, voltage gated, type II alpha subunit; sodium channel, voltage gated, type III alpha subunit; sodium channel, voltage gated, type IV alpha subunit; sodium channel, voltage gated, type IX alpha subunit; sodium channel, voltage gated, type V alpha subunit; sodium channel, voltage gated, type VIII alpha subunit; sodium channel, voltage gated, type VIII, alpha subunit; sodium channel, voltage gated, type X alpha subunit; sodium channel, voltage gated, type XI alpha subunit; sodium channel, voltage-gated, type 1, alpha polypeptide; sodium channel, voltage-gated, type I, alpha; sodium channel, voltage-gated, type I, alpha polypeptide; sodium channel, voltage-gated, type I, alpha subunit; sodium channel, voltage-gated, type II, alpha 1 polypeptide; sodium channel, voltage-gated, type II, alpha 2 polypeptide; sodium channel, voltage-gated, type II, alpha subunit; sodium channel, voltage-gated, type III, alpha polypeptide; sodium channel, voltage-gated, type IV, alpha subunit; sodium channel, voltage-gated, type IX, alpha polypeptide; sodium channel, voltage-gated, type IX, alpha subunit; sodium channel, voltage-gated, type V, alpha subunit; sodium channel, voltage-gated, type X, alpha polypeptide; sodium channel, voltage-gated, type X, alpha subunit; sodium channel, voltage-gated, type XI, alpha polypeptide; sodium channel, voltage-gated, type XI, alpha subunit; sodium channel, voltage-gated, type XII, alpha polypeptide; sodium voltage-gated channel alpha subunit 1
Gene Symbols Scn1a
Host Species Rabbit
Immunogen A synthesized peptide derived from human SCN1A(Accession P35498), corresponding to amino acid residues Q445-K495.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20265, 6323, 81574
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.