Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sodium Channel Pan Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA551019
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA551019 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA551019 Supplier Invitrogen™ Supplier No. PA551019
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCN1A Polyclonal Antibody detects endogenous levels of SCN1A protein.

Voltage-gated sodium channels (NaV) are essential for the generation of action potentials and for cell excitability. NaV channels are activated in response to depolarization and selectively allow flow of Na+ ions. To date, nine NaV alpha subunits have been cloned and named NaV1.1-NaV1.9. The NaV channels are classified into two groups according to their sensitivity to tetrodotoxin (TTX): TTX-sensitive (NaV1.1, NaV1.2, NaV1.3, NaV1.4, NaV1.6 and NaV1.7) and TTX-resistant (NaV1.5, NaV1.8 and NaV1.9). Mammalian sodium channels are heterotrimers, composed of a central, pore-forming alpha subunit and two auxiliary beta subunits. The expression of the alpha subunit isoform is developmentally regulated and tissue specific. Sodium channels in the adult central nervous system and heart contain beta 1 through beta 4 subunits, whereas sodium channels in adult skeletal muscle have only the beta 1 subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sodium Channel Pan
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SCN1A
Gene Accession No. A2APX8, O08562, O88420, O88457, P04774, P04775, P08104, P15389, P15390, P35498, P35499, Q14524, Q15858, Q62205, Q62968, Q6QIY3, Q99250, Q9ER60, Q9JJV9, Q9NY46, Q9R053, Q9UI33, Q9UQD0, Q9WTU3, Q9Y5Y9
Gene Alias 3-Feb; 6430408L10; A230052E19Rik; AI853486; alternative product; ataxia 3; B230332M13; BFIC3; BFIS3; BFNIS; brain III voltage-gated sodium channel; C630029C19Rik; cardiac tetrodotoxin-insensitive voltage-dependent sodium channel alpha subunit; CDCD2; CERIII; CIAT; CMD1E; CMPD2; CMS16; CTC-264K15.6; dmu; EIEE11; EIEE13; EIEE6; ETHA; FEB3; FEB3A; FEB3B; FEPS2; FEPS3; FHM3; GEFSP2; GEFSP7; Gm1000; HB1; HB2; HBA; HBBD; HBSC II; HBSCI; HBSCII; HH1; hNa6/Scn8a voltage-gated sodium channel; hNaN; hNE-Na; HOKPP2; hPN3; HSAN2D; HSAN7; HYKPP; HYPP; ICCD; IVF; KIAA1356; Kiaa4197; LQT3; med; mH1; mH2; microI; mKIAA4197; mnd2; mnd-2; Mu-1; Na(v)1.2; Na(V)1.4; Na(V)1.8; Na+ channel; NAC1; NAC1A; NAC2; NAC3; naCh6; NachII; NaN; NaT; Nav1.1; Nav1.2; Nav1.3; Nav1.4; Nav1.5; Nav1.5c; Nav1.6; Nav1.7; Nav1.8; NaV1.9; Nbna1; NCHVS; NENA; NE-NA; Neuroendocrine sodium channel; nmf2; nmf335; nmf58; NSS2; nur14; Peripheral nerve protein type 4; peripheral nerve sodium channel 3; Peripheral nerve sodium channel 5; peripheral sodium channel 1; PFHB1; PN1; PN3; PN4; PN5; RII/RIIA; RIIA sodium channel protein; RNSCPIIR; SCAL; SCIII; SCN; SCN1; SCN10A; SCN11A; SCN12A; SCN1A; SCN2A; SCN2A1; SCN2A2; SCN3A; SCN4A; Scn5a; SCN8A; SCN9A; ScpII; seal; Sensory neuron sodium channel; sensory neuron sodium channel 2; SFNP; skeletal muscle sodium channel alpha subunit; skeletal muscle voltage-dependent sodium channel type IV alpha subunit; skM1; SkM2; SMEI; SNS; Sns2; SNS-2; sodium channel 25; sodium channel 6; sodium channel alpha-subunit; sodium channel protein brain I subunit alpha; Sodium channel protein brain II subunit alpha; sodium channel protein brain III subunit alpha; Sodium channel protein cardiac muscle subunit alpha; sodium channel protein skeletal muscle subunit alpha; Sodium channel protein type 1 subunit alpha; sodium channel protein type 10 subunit alpha; Sodium channel protein type 11 subunit alpha; sodium channel protein type 2 subunit alpha; Sodium channel protein type 3 subunit alpha; sodium channel protein type 4 subunit alpha; Sodium channel protein type 5 subunit alpha; sodium channel protein type 8 subunit alpha; Sodium channel protein type 9 subunit alpha; sodium channel protein type I subunit alpha; Sodium channel protein type II subunit alpha; sodium channel protein type III subunit alpha; Sodium channel protein type IV subunit alpha; sodium channel protein type IX subunit alpha; Sodium channel protein type V subunit alpha; sodium channel protein type VIII subunit alpha; Sodium channel protein type X subunit alpha; sodium channel protein type XI subunit alpha; sodium channel protein, brain I alpha subunit; sodium channel protein, brain I subunit alpha; sodium channel protein, brain II subunit alpha; sodium channel protein, brain III subunit alpha; sodium channel protein, brain type 2 alpha subunit; sodium channel type X alpha polypeptide; sodium channel voltage gated type 1 alpha subunit; sodium channel voltage-gated type 11 alpha polypeptide; sodium channel voltage-gated type 4 alpha polypeptide; sodium channel voltage-gated type I alpha polypeptide; sodium channel voltage-gated type II alpha polypeptide; Sodium channel voltage-gated type IV alpha polypeptide; sodium channel voltage-gated type V alpha polypeptide; sodium channel voltage-gated type VIII alpha polypeptide; sodium channel voltage-gated type X alpha polypeptide; sodium channel voltage-gated type XI alpha polypeptide; sodium channel, voltage gated, type I alpha subunit; sodium channel, voltage gated, type II alpha subunit; sodium channel, voltage gated, type III alpha subunit; sodium channel, voltage gated, type IV alpha subunit; sodium channel, voltage gated, type IX alpha subunit; sodium channel, voltage gated, type V alpha subunit; sodium channel, voltage gated, type VIII alpha subunit; sodium channel, voltage gated, type VIII, alpha subunit; sodium channel, voltage gated, type X alpha subunit; sodium channel, voltage gated, type XI alpha subunit
Gene Symbols SCN10A, Scn11a, SCN1A, SCN2A, SCN3A, Scn4a, Scn5a, Scn8a, SCN9A
Host Species Rabbit
Immunogen Synthesized peptide derived from the Internal region of human Na+ CP-pan.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110876, 110880, 11280, 20264, 20265, 20269, 20271, 20273, 20274, 24046, 24766, 25665, 25722, 29571, 29701, 29710, 497770, 6323, 6326, 6328, 6329, 6331, 6334, 6335, 6336, 78956, 81574
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.