Learn More
Invitrogen™ SCN1B Polyclonal Antibody

Description
This antibody is predicted to react with bovine, rat and rabbit based on sequence homology.
Specifications
Specifications
| Antigen | SCN1B |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.48 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Scn1b |
| Gene Accession No. | P97952, Q07699 |
| Gene Alias | ATFB13; BRGDA5; GEFSP1; Scn1b; sodium channel beta 1-subunit; sodium channel beta-1 subunit; sodium channel subunit beta-1; sodium channel voltage-gated type 1 beta polypeptide; sodium channel voltage-gated type I beta polypeptide; sodium channel, voltage gated, type I beta subunit; sodium channel, voltage-gated, type 1, beta polypeptide; sodium channel, voltage-gated, type I, beta; sodium channel, voltage-gated, type I, beta polypeptide; sodium channel, voltage-gated, type I, beta subunit; sodium voltage-gated channel beta subunit 1 |
| Gene Symbols | Scn1b |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.