Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCN1B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987429
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7429 Supplier Bioss Supplier No. BS6687R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Voltage-gated sodium channels are heteromeric proteins that function in the generation and propagation of action potentials in muscle and neuronal cells. They are composed of one alpha and two beta subunits, where the alpha subunit provides channel activity and the beta-1 subunit modulates the kinetics of channel inactivation. SCN1B encodes a sodium channel beta-1 subunit. Mutations in this gene result in generalized epilepsy with febrile seizures plus, Brugada syndrome 5, and defects in cardiac conduction. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen SCN1B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Scn1b
Gene Accession No. Q07699
Gene Alias ATFB13; BOS_17576; BRGDA5; GEFSP1; Scn1b; sodium channel beta 1-subunit; sodium channel beta-1 subunit; Sodium channel subunit beta-1; sodium channel voltage-gated type 1 beta polypeptide; sodium channel voltage-gated type I beta polypeptide; sodium channel, voltage gated, type I beta subunit; sodium channel, voltage-gated, type 1, beta polypeptide; sodium channel, voltage-gated, type I, beta; sodium channel, voltage-gated, type I, beta polypeptide; sodium channel, voltage-gated, type I, beta subunit; sodium voltage-gated channel beta subunit 1
Gene Symbols Scn1b
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SCN1B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6324
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.