Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ SCN2A, Mouse, Unlabeled, Clone: 5H10.2,

Catalog No. MABN1586MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1586MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1586MI Supplier MilliporeSigma™ Supplier No. MABN1586
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sodium channel protein type 2 subunit alpha (UniProt Q99250; also known as HBSC II, Sodium channel protein brain II subunit alpha, Sodium channel protein type II subunit alpha, Voltage-gated sodium channel subunit alpha Nav1.2) is encoded by the SCN2A (also known as EIEE11, BFIS3, NAC2, SCN2A1, SCN2A2) gene (Gene ID 6326) in human. Sodium channels are integral plasma membrane proteins that conduct Na+ transport across the plasma membrane. They are classified as either voltage-gated or ligand-gated based on the nature of the triggering mechanism for channel activation. Voltage-gated sodium channels (VGSCs) are also called voltage-sensitive, voltage-dependent, or Nav channels. VGSCs normally consist of an alpha subunit that forms the ion-conducting pore and one to two beta subunits that have several functions including channel gating modulation, although the expression of the alpha subunit alone is sufficient to produce a functional channel. There exist nine known alpha subunits (Nav1.1 through Nav1.5 encoded by SCN1A through SCN5A and Nav1.6 through Nav1.9 encoded by SCN8A through SCN11A) and four beta subunits (Navbeta1 through Navbeta4 encoded by SCN1B through SCN4B). Beta 1 and beta 3 interact with the alpha subunit non-covalently, whereas beta 2 and beta 4 associate with alpha via disulfide bond. Nav1.2 is a multi-pass membrane protein with 24 membrane regions, having both its N- and C-terminal ends at the cytoplasmic side (a.a. 1-124 and 1777-2005).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCN2A
Applications Western Blot
Classification Monoclonal
Clone 5H10.2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene SCN2A, EIEE11, BFIS3, NAC2, SCN2A1, SCN2A2
Gene Accession No. Q99250
Host Species Mouse
Immunogen GST-tagged recombinant human SCN2A C-terminal cytoplasmic domain fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001035232
Test Specificity Clone 5H10.2 targets an epitope in the C-terminal cytoplasmic domain.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.