Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scn2a1 Mouse anti-Rat, Alkaline Phosphatase, Clone: CJ6, Abnova™

Catalog No. 89339011 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-011 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-011 Supplier Abnova Corporation Supplier No. MAB19726

Mouse Monoclonal Antibody.

Scn2a1 Monoclonal antibody specifically detects Scn2a1 in Rat samples. It is validated for WB, IHC-P.

Specifications

Antigen Scn2a1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CJ6
Conjugate Alkaline Phosphatase (AP)
Description sodium channel, voltage-gated, type II, alpha 1
Gene Scn2a1
Gene Alias NachII, Nav1.2, RII/RIIA, RNSCPIIR, SCN, SCN2A, Scn2a2, ScpII
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1882-2005 at C-terminus of rat Scn2a1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 24766
Target Species Rat
Content And Storage Store at 4°C
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.