Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Scn2b Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300207MFV500
Description
Scn2b Polyclonal antibody specifically detects Scn2b in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
Scn2b | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
neuronal voltage-gated sodium channel beta 2 subunit, sodium channel beta 2 subunit, sodium channel subunit beta-2, sodium channel, voltage-gated, type II, beta, sodium channel, voltage-gated, type II, beta polypeptide | |
Produced in rabbits immunized with purified, recombinant Human Scn2b (Accession#: O60939; Met1-Ala159) | |
0.1 mL | |
Neuroscience | |
6327 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction