Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCN2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101892
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101892 Supplier Invitrogen™ Supplier No. PA5101892
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SCN2B.

Scn2b is a member of voltage-gated sodium channels (NaV) that are responsible for action potential initiation and propagation in excitable cells, including nerve, muscle, and neuroendocrine cell types. Scn2b are also expressed at low levels in non-excitable cells, where their physiological role is unclear. Scn2b is crucial in the assembly, expression, and functional modulation of the heterotrimeric complex of the sodium channel. In Scn2b, the subunit beta-2 causes an increase in the plasma membrane surface area and in its folding into microvilli. Also, Scn2b interacts with TNR (Tenascin R-precursor) and may play a crucial role in clustering and regulation of activity of sodium channels at nodes of Ranvier. The protein encoded by the Scn2b gene is the beta 2 subunit of the type II voltage-gated sodium channel. Scn2b protein is involved in cell-cell adhesion and cell migration. Defects in the Scn2b gene can be a cause of Brugada Syndrome, atrial fibrillation, or sudden infant death syndrome. Diseases associated with SCN2B include Atrial Fibrillation, Familial, 14 and Scn2b-Related Familial Atrial Fibrillation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCN2B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SCN2B
Gene Accession No. O60939, P54900, Q56A07
Gene Alias 2810451E09Rik; AI840361; ATFB14; Gm183; neuronal voltage-gated sodium channel beta 2 subunit; Scn2b; SCNB2; Sodium channel beta 2; sodium channel subunit beta-2; sodium channel, voltage gated, type II beta subunit; sodium channel, voltage-gated, type II, beta; sodium channel, voltage-gated, type II, beta polypeptide; sodium channel, voltage-gated, type II, beta subunit; sodium voltage-gated channel beta subunit 2; UNQ326/PRO386
Gene Symbols SCN2B
Host Species Rabbit
Immunogen A synthesized peptide derived from human SCN2B(Accession O60939), corresponding to amino acid residues Q70-E120.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25349, 6327, 72821
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.