Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCN3A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587615
Description
Immunogen sequence: RNKCLQWPPS DSAFETNTTS YFNGTMDSNG TFVNVTMSTF NWKDYIGDDS HFYVLDGQKD PLLCGNGSDA GQCPEGYICV KAGRNPNYGY TSFDTFSWAF LSLFRLMTQD YWENLYQLTL RAAGKTY; Positive Samples: 293T, HT-1080; Cellular Location: Membrane, Multi-pass membrane protein.
Mediates the voltage-dependent sodium ion permeability of excitable membranes. Assuming opened or closed conformations in response to the voltage difference across the membrane, the protein forms a sodium-selective channel through which Na+ ions may pass in accordance with their electrochemical gradient.
Specifications
SCN3A | |
Polyclonal | |
Unconjugated | |
Scn3a | |
BIII; brain III voltage-gated sodium channel; Gm1000; KIAA1356; NAC3; Nav1.3; SCIII; Scn2a; Scn3a; Sodium channel protein brain III subunit alpha; sodium channel protein type 3 subunit alpha; Sodium channel protein type III subunit alpha; sodium channel protein, brain III subunit alpha; sodium channel, voltage gated, type III alpha subunit; sodium channel, voltage-gated, type 3, alpha subunit; sodium channel, voltage-gated, type III, alpha; sodium channel, voltage-gated, type III, alpha polypeptide; sodium channel, voltage-gated, type III, alpha subunit; sodium voltage-gated channel alpha subunit 3; voltage-gated sodium channel subtype III; Voltage-gated sodium channel subunit alpha Nav1.3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
6328 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.99 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NY46 | |
Scn3a | |
Recombinant fusion protein containing a sequence corresponding to amino acids 274-400 of human SCN3A (NP_0088533). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction