Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCN3A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA577724
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Mediates the voltage-dependent sodium ion permeability of excitable membranes. Assuming opened or closed conformations in response to the voltage difference across the membrane, the protein forms a sodium-selective channel through which Na+ ions may pass in accordance with their electrochemical gradient.
Specifications
SCN3A | |
Polyclonal | |
Unconjugated | |
SCN3A | |
Brain Type III | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20269, 497770, 6328 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P08104, Q9NY46 | |
SCN3A | |
Peptide (C)HLEGNHRADGDRFP, corresponding to amino acid residues 511-524 rat Nav 1.3 | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction