Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCN3A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981967
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1967 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1967 Supplier Bioss Supplier No. BS12133R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mediates the voltage-dependent sodium ion permeability of excitable membranes. Assuming opened or closed conformations in response to the voltage difference across the membrane, the protein forms a sodium-selective channel through which Na+ ions may pass in accordance with their electrochemical gradient.

Specifications

Antigen SCN3A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Scn3a
Gene Accession No. Q9NY46
Gene Alias BIII; brain III voltage-gated sodium channel; Gm1000; KIAA1356; NAC3; Nav1.3; SCIII; Scn2a; SCN3A; Sodium channel protein brain III subunit alpha; Sodium channel protein type 3 subunit alpha; Sodium channel protein type III subunit alpha; sodium channel protein, brain III subunit alpha; sodium channel, voltage gated, type III alpha subunit; sodium channel, voltage-gated, type 3, alpha subunit; sodium channel, voltage-gated, type III, alpha; sodium channel, voltage-gated, type III, alpha polypeptide; sodium channel, voltage-gated, type III, alpha subunit; sodium voltage-gated channel alpha subunit 3; Voltage-gated sodium channel subtype III; Voltage-gated sodium channel subunit alpha Nav1.3
Gene Symbols Scn3a
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SCN3A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6328
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.