Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCN3B Antibody (S396-29), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259318UV
Description
SCN3B Monoclonal specifically detects SCN3B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
SCN3B | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SCN3B | |
Fusion protein amino acids 1-215 (full-length) of rat NavBeta3 | |
100 μL | |
55800 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
S396-29 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
HSA243396, KIAA1158, SCNB3, sodium channel subunit beta-3, sodium channel, voltage-gated, type III, beta, voltage-gated sodium channel beta-3 subunit | |
Mouse | |
Protein G purified | |
Primary | |
Detects 40kDa. Does not cross-reat with NavBeta1, Navbeta2, or Navbeta4. | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction