Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCN3B Monoclonal Antibody (N396/29), PerCP
GREENER_CHOICE

Catalog No. PIMA545739
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545739 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545739 Supplier Invitrogen™ Supplier No. MA545739
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:100 dilution of MA5-45739 was sufficient for detection of NavBeta3 in 20 μg of mouse brain lysate by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 40kDa. Does not cross-reat with NavBeta1, Navbeta2, or Navbeta4. This antibody was formerly sold as clone S396-29.

Voltage-gated sodium channels are transmembrane glycoprotein complexes composed of a large alpha subunit and one or more regulatory beta subunits. They are responsible for the generation and propagation of action potentials in neurons and muscle. This gene encodes one member of the sodium channel beta subunit gene family, and influences the inactivation kinetics of the sodium channel. Two alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCN3B
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N396/29
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene Scn3b
Gene Accession No. Q8BHK2, Q9JK00, Q9NY72
Gene Alias 1110001K16Rik; 4833414B02Rik; ATFB16; brain and heart sodium channel beta 3 subunit; BRGDA7; HSA243396; KIAA1158; LOC540925 protein; Scn3b; Scnb3; sodium channel beta 3 subunit; sodium channel subunit beta-3; sodium channel, voltage gated, type III beta subunit; sodium channel, voltage-gated, type III, beta; sodium channel, voltage-gated, type III, beta subunit; sodium voltage-gated channel beta subunit 3; voltage-gated sodium channel beta-3 subunit
Gene Symbols Scn3b
Host Species Mouse
Immunogen Fusion protein amino acids 1-215 (full-length) of rat NavBeta3.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235281, 245956, 55800
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.