Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scn3b Mouse anti-Human, Mouse, Rat, ATTO 390, Clone: S396-29, Abnova™

Catalog No. 89338960 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-960 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-960 Supplier Abnova Corporation Supplier No. MAB19456
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

Scn3b Monoclonal antibody specifically detects Scn3b in Human, Mouse, Rat samples. It is validated for ICC, WB-Ti, IF.

Specifications

Antigen Scn3b
Applications Immunocytochemistry, Western Blot, Immunofluorescence
Classification Monoclonal
Clone S396-29
Conjugate Atto 390
Description sodium channel, voltage-gated, type III, beta
Gene Scn3b
Gene Alias Scnb3
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-215 at C-terminus region of rat Scn3b
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 245956
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.