Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scn3b Mouse anti-Mouse, Rat, APC, Clone:S396-29, Abnova™

Catalog No. 89334223 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-223 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-223 Supplier Abnova Corporation Supplier No. MAB17355
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full length recombinant rat Scn3b.

Specifications

Antigen Scn3b
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone S396-29
Conjugate APC
Description sodium channel, voltage-gated, type III, beta
Dilution Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Scn3b
Gene Alias Scnb3
Host Species Mouse
Immunogen Recombinant protein corresponding to full length rat Scn3b.
Quantity 100 μg
Gene ID (Entrez) 245956
Target Species Mouse, Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.