Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCN4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553579
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553579 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553579 Supplier Invitrogen™ Supplier No. PA553579
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLPCTFSSCF GFEDLHFRWT YNSSDAFKIL IEGTVKNEKS DPKVTLKDDD RITLVGSTKE KMNNISIVLR DLEFSDTGKY TCHVKNPKEN NLQHHATIFL QVV Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 76%.

The SCN4B protein modulates channel gating kinetics. Causes negative shifts in the voltage dependence of activation of certain alpha sodium channels, but does not affect the voltage dependence of inactivation. The protein has been found to be expressed at a high level in dorsal root ganglia, at a lower level in brain, spinal cord, skeletal muscle and heart. Defects in SCN4B are the cause of long QT syndrome type 10 (LQT10) [MIM:611819]. Long QT syndromes are heart disorders characterized by a prolonged QT interval on the ECG and polymorphic ventricular arrhythmias. They cause syncope and sudden death in response to exercise or emotional stress. They can present with a sentinel event of sudden cardiac death in infancy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCN4B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SCN4B
Gene Accession No. Q8IWT1
Gene Alias ATFB17; Gm1471; LQT10; Navbeta4; Scn4b; sodium channel accessory subunit; sodium channel beta 4 subunit; sodium channel subunit beta-4; sodium channel, type 4, beta subunit; sodium channel, type IV, beta; sodium channel, type IV, beta polypeptide; sodium channel, voltage gated, type IV beta subunit; sodium channel, voltage-gated, type IV, beta subunit; sodium voltage-gated channel beta subunit 4; voltage gated sodium channel beta 4 subunit
Gene Symbols SCN4B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SCN4B. Recombinant protein control fragment (Product #RP-90488).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6330
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.