Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCN4B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115872
Description
Antibody detects endogenous levels of total SCN4B.
The SCN4B protein modulates channel gating kinetics. Causes negative shifts in the voltage dependence of activation of certain alpha sodium channels, but does not affect the voltage dependence of inactivation. The protein has been found to be expressed at a high level in dorsal root ganglia, at a lower level in brain, spinal cord, skeletal muscle and heart. Defects in SCN4B are the cause of long QT syndrome type 10 (LQT10) [MIM:611819]. Long QT syndromes are heart disorders characterized by a prolonged QT interval on the ECG and polymorphic ventricular arrhythmias. They cause syncope and sudden death in response to exercise or emotional stress. They can present with a sentinel event of sudden cardiac death in infancy.
Specifications
SCN4B | |
Polyclonal | |
Unconjugated | |
SCN4B | |
ATFB17; Gm1471; LQT10; Navbeta4; Scn4b; sodium channel accessory subunit; sodium channel beta 4 subunit; sodium channel subunit beta-4; sodium channel, type 4, beta subunit; sodium channel, type IV, beta; sodium channel, type IV, beta polypeptide; sodium channel, voltage gated, type IV beta subunit; sodium channel, voltage-gated, type IV, beta subunit; sodium voltage-gated channel beta subunit 4; voltage gated sodium channel beta 4 subunit | |
Rabbit | |
Affinity chromatography | |
RUO | |
315611, 399548, 6330 | |
-20°C | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q7M729, Q7M730, Q8IWT1 | |
SCN4B | |
A synthesized peptide derived from human SCN4B(Accession Q8IWT1), corresponding to amino acid residues K184-V228. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction