Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCN5A Monoclonal Antibody (4G8:1G7)
GREENER_CHOICE

Catalog No. PIMA524779
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524779 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524779 Supplier Invitrogen™ Supplier No. MA524779
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: VSENIKLGNLSALR Purified by Thiophilic Chromatography Immunogen sequence: VSENIKLGNL SALR.

Scn5a is an integral membrane protein and tetrodotoxin-resistant voltage-gated sodium channel subunit. Scn5a is found primarily in cardiac muscle and is responsible for the initial upstroke of the action potential in an electrocardiogram. Defects in Scn5a are a cause of long QT syndrome type 3 (LQT3), an autosomal dominant cardiac disease. Alternative splicing results in several transcript variants encoding different isoforms of Scn5a. Voltage-gated sodium channesl (VGSCs) mediate regenerative cell membrane depolarization and conduction of electrical signalling in nerves and muscles. Expression of Scn5a is also detected in lymphocytes, glia, and fibroblasts. Further, Scn5a mediates the voltage-dependent sodium ion permeability of excitable membranes. Assuming opened or closed conformations in response to the voltage difference across the membrane, Scn5a forms a sodium-selective channel through which Na(+) ions may pass in accordance with their electrochemical gradient. Functionally, Scn5a is responsible for the initial upstroke of the action potential and the channel inactivation is regulated by intracellular calcium levels. Voltage-gated sodium channels (NaV) are responsible for action potential initiation and propagation in excitable cells, including nerve, muscle, and neuroendocrine cell types. Scn5a are also expressed at low levels in non-excitable cells, where their physiological role is unclear. Diseases associated with SCN5A include Long Qt Syndrome-3 and Brugada Syndrome 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCN5A
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 4G8:1G7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SCN5A
Gene Accession No. Q14524
Gene Alias cardiac tetrodotoxin-insensitive voltage-dependent sodium channel alpha subunit; CDCD2; CDCD2VF1; CIN5; CMD1E; CMPD2; HB1; HB2; HBBD; HH1; HH1CMD1E; ICCD; IVF; LQT3; LQT3SSS1CMPD2; mH1; Nav1.5; Nav1.5c; PFHB1; RATRSKM2X; RH1; RSkM2; RSKM2X; SCAL; Scn2x; SCN5A; SkM1; SkM2; sodium channel protein cardiac muscle subunit alpha; Sodium channel protein type 5 subunit alpha; sodium channel protein type V subunit alpha; sodium channel voltage-gated type V alpha polypeptide; sodium channel, voltage gated, type V alpha subunit; sodium channel, voltage-gated, type 5, alpha subunit; sodium channel, voltage-gated, type V, alpha; sodium channel, voltage-gated, type V, alpha polypeptide; sodium channel, voltage-gated, type V, alpha subunit; sodium voltage-gated channel alpha subunit 5; SSS1; VF1; voltage-gated sodium channel Nav1.5c; voltage-gated sodium channel subunit alpha Nav1.5
Gene Symbols SCN5A
Host Species Mouse
Immunogen Peptide (14mer) from an extracellular domain of Nav1.5.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6331
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.