Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCN7A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550138 Supplier Invitrogen™ Supplier No. PA550138
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total SCN7A protein.

Mediates the voltage-dependent sodium ion permeability of excitable membranes. Assuming opened or closed conformations in response to the voltage difference across the membrane, the protein forms a sodium-selective channel through which Na+ ions may pass in accordance with their electrochemical gradient. George A.L. Jr., Proc. Natl. Acad. Sci. U.S.A. 89:4893-4897(1992).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCN7A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SCN7A
Gene Accession No. Q01118
Gene Alias 1110034K09Rik; AI642000; LOW QUALITY PROTEIN: sodium channel protein type 7 subunit alpha; NaG; Na-G; Nav2; Nav2.1; Nav2.2; Nav2.3; Nax; putative voltage-gated sodium channel subunit alpha Nax; Scn6a; SCN7A; Sodium channel protein cardiac and skeletal muscle subunit alpha; sodium channel protein type 7 subunit alpha; Sodium channel protein type VII subunit alpha; sodium channel voltage-gated type VI alpha polypeptide; sodium channel voltage-gated type VII alpha; sodium channel, voltage gated, type VII alpha subunit; sodium channel, voltage-gated, type 6, alpha polypeptide; sodium channel, voltage-gated, type VI, alpha polypeptide; sodium channel, voltage-gated, type VII, alpha; sodium channel, voltage-gated, type VII, alpha subunit; sodium voltage-gated channel alpha subunit 7; voltage-dependent sodium channel alpha subunit
Gene Symbols SCN7A
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human SCN7A.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6332
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.