Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCN7A Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12127R

Description
Mediates the voltage-dependent sodium ion permeability of excitable membranes. Assuming opened or closed conformations in response to the voltage difference across the membrane, the protein forms a sodium-selective channel through which Na+ ions may pass in accordance with their electrochemical gradient. George A.L. Jr., Proc. Natl. Acad. Sci. U.S.A. 89:4893-4897(1992).Specifications
SCN7A | |
Polyclonal | |
Unconjugated | |
SCN7A | |
1110034K09Rik; AI642000; LOW QUALITY PROTEIN: sodium channel protein type 7 subunit alpha; NaG; Na-G; Nav2; Nav2.1; Nav2.2; Nav2.3; Nax; Putative voltage-gated sodium channel subunit alpha Nax; SCN6A; SCN7A; Sodium channel protein cardiac and skeletal muscle subunit alpha; Sodium channel protein type 7 subunit alpha; Sodium channel protein type VII subunit alpha; sodium channel voltage-gated type VI alpha polypeptide; sodium channel voltage-gated type VII alpha; sodium channel, voltage gated, type VII alpha subunit; sodium channel, voltage-gated, type 6, alpha polypeptide; sodium channel, voltage-gated, type VI, alpha polypeptide; sodium channel, voltage-gated, type VII, alpha; sodium channel, voltage-gated, type VII, alpha subunit; sodium voltage-gated channel alpha subunit 7; voltage-dependent sodium channel alpha subunit | |
Rabbit | |
Protein A | |
RUO | |
6332 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q01118 | |
SCN7A | |
KLH conjugated synthetic peptide derived from human SCN7A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction