Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCN8A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113521
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113521 Supplier Invitrogen™ Supplier No. PA5113521
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SCN8A Polyclonal Antibody for Western Blot, IHC (P), ELISA

The sodium channel protein type 8 subunit alpha (Na+ CP type VIII alpha ) is a multi-pass, transmembrane protein that mediates the sodium ion permeability of excitable membranes. The three glycoproteins that comprise the voltage-gated sodium channel proteins include a pore-forming alpha subunit, a noncovalently associated beta1 subunit and a disulfide-linked beta2 subunit. The two beta subunits regulate the level of channel expression, modulate gating and function as cell adhesion molecules for cellular aggregation and cytoskeleton interaction. The alpha subunits of sodium channels type I and III are predominantly expressed in neuronal cell bodies and proximal processes, while type II alpha subunits are more abundant along axons. Sodium channels are important for rapid signal transduction but also play a significant role in neuronal development. Defects of the SCN8A gene have exhibited detrimental effects on the growth of secondary motoneurons. Loss of SCN8A expression will result in progressive paralysis and early death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCN8A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Scn8a
Gene Accession No. O88420, Q9UQD0
Gene Alias AI853486; ataxia 3; C630029C19Rik; CERIII; CIAT; dmu; EIEE13; hNa6/Scn8a voltage-gated sodium channel; MED; mnd2; mnd-2; Na+ channel; naCh6; Nav1.6; Nbna1; nmf2; nmf335; nmf58; nur14; peripheral nerve protein type 4; PN4; SCN8A; Scn8a (PN4); seal; Sodium channel 6; sodium channel protein type 8 subunit alpha; Sodium channel protein type VIII subunit alpha; sodium channel voltage-gated type VIII alpha polypeptide; sodium channel, voltage gated, type VIII alpha subunit; sodium channel, voltage gated, type VIII, alpha subunit; sodium channel, voltage-gated, type 8, alpha polypeptide; sodium channel, voltage-gated, type 8, alpha subunit; sodium channel, voltage-gated, type VIII, alpha; sodium channel, voltage-gated, type VIII, alpha polypeptide; sodium voltage-gated channel alpha subunit 8; voltage-gated sodium channel subunit alpha Nav1.6; voltage-gated sodium channel type VIII alpha protein
Gene Symbols Scn8a
Host Species Rabbit
Immunogen Recombinant Human Sodium channel protein type 8 subunit alpha protein (1350-1440AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29710, 6334
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.