Learn More
Invitrogen™ SCNM1 Monoclonal Antibody (1E10)

Description
Peptide Sequence: MSFKREGDDW SQLNVLKKRR VGDLLASYIP EDEALMLRDG RFACAICPHR PVLDTLAMLT AHRAGKKHLS SLQLFYGKKQ PGKERKQNPK HQNELRREET KAEAPLLTQT RLITQSALHR APHYNSCCRR KYRPEAPGPS VSLSPMPPSE VKLQSGKISR EPEPAAGPQA EESATVSAPA PMSPTRRRAL DHYLTLRSSG WIPDGRGRWV KDENVEFDSD EEEPPDLPLD.
Specifications
Specifications
| Antigen | SCNM1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1E10 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SCNM1 |
| Gene Accession No. | Q9BWG6 |
| Gene Alias | 3110001I17Rik; Scnm1; Scnm1-ps; Sodium channel modifier 1; sodium channel modifier 1, pseudogene |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.