Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCNM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545736
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545736 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545736 Supplier Invitrogen™ Supplier No. PA545736
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EVKLQSGKIS REPEPAAGPQ AEESATVSAP APMSPTRRRA LDHYLTLRSS Sequence homology: Horse: 79%; Human: 100%; Rabbit: 79%.

SCNM1 is a zinc finger protein and putative splicing factor. In mice, Scnm1 modifies phenotypic expression of Scn8a mutations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCNM1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SCNM1
Gene Accession No. Q9BWG6
Gene Alias 3110001I17Rik; Scnm1; Scnm1-ps; Sodium channel modifier 1; sodium channel modifier 1, pseudogene
Gene Symbols SCNM1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human SCNM1 (aa 150-199).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79005
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.